Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040156.1 | - | 5 - 6 | 936 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |