Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138655.1 | NP_619596.1 | 6 - 7 | 995 | 139 | |
GenBank mRNA | AF417581.1 | - | 6 - 7 | 995 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |