Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133655.2 | NP_598416.1 | 4 - 5 | 602 | 73 | |
GenBank mRNA | AK136370.1 | - | 4 - 5 | 566 | 73 | |
AK166521.1 | - | 4 - 5 | 568 | 73 | ||
AK170741.1 | - | 4 - 5 | 567 | 73 | ||
AK182212.1 | - | 1 - 2 | 74 | 73 | ||
AK185682.1 | - | 1 - 2 | 74 | 73 | ||
AK211761.1 | - | 1 - 2 | 74 | 73 | ||
BC011433.1 | - | 4 - 5 | 557 | 2811801 | 73 | |
X59047.1 | - | 4 - 5 | 322 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |