Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252543.1 | NP_001239472.1 | 9 - 10 | 1230 | 143 | |
NM_001252544.1 | NP_001239473.1 | 9 - 10 | 1223 | 143 | ||
NM_001252545.1 | NP_001239474.1 | 9 - 10 | 1212 | 143 | ||
NM_001281816.1 | NP_001268745.1 | 6 - 7 | 852 | 143 | ||
NM_019787.4 | NP_062761.2 | 9 - 10 | 1241 | 143 | ||
GenBank mRNA | AF200326.1 | - | 8 - 9 | 1113 | 143 | |
AK014597.1 | - | 9 - 10 | 1191 | 143 | ||
AK029661.1 | - | 6 - 7 | 853 | 143 | ||
AK139729.1 | - | 9 - 10 | 1156 | 143 | ||
AK146083.1 | - | 9 - 10 | 1190 | 143 | ||
AK149693.1 | - | 9 - 10 | 1170 | 143 | ||
AK153214.1 | - | 9 - 10 | 1197 | 143 | ||
AK158398.1 | - | 9 - 10 | 1208 | 143 | ||
AK166330.1 | - | 9 - 10 | 1173 | 143 | ||
AK167382.1 | - | 9 - 10 | 1179 | 143 | ||
AK167732.1 | - | 9 - 10 | 1159 | 143 | ||
AK171597.1 | - | 9 - 10 | 1171 | 143 | ||
AK209568.1 | - | 2 - 3 | 235 | 143 | ||
AK220080.1 | - | 2 - 3 | 234 | 144 | ||
BC005464.1 | - | 9 - 10 | 1166 | 3591420 | 143 | |
BC011160.1 | - | 9 - 10 | 1193 | 4024353 | 143 | |
BU614902.1 | - | 5 - 6 | 570 | 143 | ||
CF724836.1 | - | 3 - 4 | 420 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |