Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146010.1 | NP_001139482.1 | 10 - 11 | 1164 | 68 | |
GenBank mRNA | AK140322.1 | - | 10 - 11 | 1143 | 68 | |
AK141949.1 | - | 10 - 11 | 1164 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |