Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183208.3 | NP_899031.2 | 23 - 24 | 3722 | 58 | |
GenBank mRNA | BC057691.1 | - | 13 - 14 | 1959 | 58 | |
BC058646.1 | - | 23 - 24 | 3622 | 6405777 | 58 | |
BC065120.1 | - | 23 - 24 | 3615 | 6856060 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |