Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100021798.mm.1 |
NM_172430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021798.mm.1 |
NM_172430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021798.mm.1 |
TC0100002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021802.mm.1 |
NM_172430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021802.mm.1 |
NM_172430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021802.mm.1 |
TC0100002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021803.mm.1 |
NM_172430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021803.mm.1 |
NM_172430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021803.mm.1 |
TC0100002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021799.mm.1 |
NM_172430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021799.mm.1 |
NM_172430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021799.mm.1 |
TC0100002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021805.mm.1 |
NM_172430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021805.mm.1 |
NM_172430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021805.mm.1 |
TC0100002685.mm.1 |
MTA TC |