Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031863.3 | NP_114069.1 | 4 - 5 | 487 | 154 | |
GenBank mRNA | AK011998.1 | - | 4 - 5 | 473 | 154 | |
AK012179.1 | - | 4 - 5 | 488 | 154 | ||
AK020040.1 | - | 4 - 5 | 489 | 154 | ||
AK076094.1 | - | 4 - 5 | 465 | 154 | ||
AK157569.1 | - | 4 - 5 | 512 | 154 | ||
BC018524.1 | - | 4 - 5 | 420 | 3588374 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |