Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168273.1 | NP_001161745.1 | 16 - 17 | 2056 | 68 | |
NM_172596.2 | NP_766184.2 | 15 - 16 | 2284 | 68 | ||
GenBank mRNA | AK036519.1 | - | 15 - 16 | 2284 | 68 | |
AK122199.1 | - | 16 - 17 | 2034 | 68 | ||
AK132090.1 | - | 10 - 11 | 1311 | 68 | ||
BC027157.1 | - | 2 - 3 | 210 | 68 | ||
BC027775.1 | - | 6 - 7 | 549 | 68 | ||
BC040370.1 | - | 15 - 16 | 2262 | 4239204 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |