Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009502.4 | NP_033528.3 | 9 - 10 | 1260 | 70 | |
GenBank mRNA | AK077850.1 | - | 9 - 10 | 1296 | 70 | |
BC008520.1 | - | 9 - 10 | 1260 | 3495710 | 70 | |
L18880.1 | - | 9 - 10 | 1189 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |