Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205241.1 | NP_001192170.1 | 9 - 10 | 1652 | 130 | |
NM_017479.3 | NP_059507.2 | 9 - 10 | 1776 | 130 | ||
GenBank mRNA | AF222800.1 | - | 9 - 10 | 1692 | 130 | |
AK045188.1 | - | 9 - 10 | 2115 | 130 | ||
AK147509.1 | - | 9 - 10 | 1653 | 130 | ||
BC095974.1 | - | 8 - 9 | 1429 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |