Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013767.2 | NP_001013789.1 | 20 - 21 | 2107 | 67 | |
GenBank mRNA | AY578330.1 | - | 20 - 21 | 2107 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |