Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162864.1 | NP_001156336.1 | 7 - 8 | 947 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |