Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081335.2 | NP_001074804.2 | 15 - 16 | 3631 | 76 | |
GenBank mRNA | AK122351.1 | - | 12 - 13 | 2622 | 76 | |
AK147675.1 | - | 14 - 15 | 3886 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |