Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008598.2 | NP_032624.1 | 1 - 2 | 73 | 61 | |
GenBank mRNA | AK090389.1 | - | 61 | 61 | ||
BC031888.1 | - | 1 - 2 | 42 | 5372377 | 61 | |
BC096557.1 | - | 1 - 2 | 76 | 1366095 | 61 | |
BY344992.1 | - | 1 - 2 | 81 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |