Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198608.2 | NP_941010.2 | 9 - 10 | 1297 | 60 | |
GenBank mRNA | AK146958.1 | - | 9 - 10 | 1297 | 60 | |
AK167771.1 | - | 9 - 10 | 1292 | 60 | ||
AK173139.1 | - | 5 - 6 | 563 | 60 | ||
BC079844.1 | - | 9 - 10 | 1269 | 60 | ||
BC113172.1 | - | 9 - 10 | 1293 | 40060953 | 60 | |
BC113779.1 | - | 9 - 10 | 1293 | 40060952 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |