Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081238.2 | NP_001074707.1 | 6 - 7 | 2521 | 66 | |
NM_001289842.1 | NP_001276771.1 | 6 - 7 | 2521 | 66 | ||
NM_001289843.1 | NP_001276772.1 | 5 - 6 | 2427 | 66 | ||
NM_001289844.1 | NP_001276773.1 | 6 - 7 | 1432 | 66 | ||
NM_001289845.1 | NP_001276774.1 | 3 - 4 | 361 | 66 | ||
GenBank mRNA | AK154227.1 | - | 6 - 7 | 2493 | 66 | |
AK156701.1 | - | 6 - 7 | 2440 | 66 | ||
AK157382.1 | - | 6 - 7 | 2440 | 66 | ||
BC044748.2 | - | 2 - 3 | 126 | 66 | ||
BC055302.1 | - | 3 - 4 | 324 | 66 | ||
BC066139.1 | - | 5 - 6 | 2369 | 66 | ||
BC066140.1 | - | 6 - 7 | 1330 | 30459312 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |