Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC141310.1 | - | 3 - 4 | 726 | 9055922 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |