Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198029.2 | NP_932146.2 | 12 - 13 | 2327 | 119 | |
GenBank mRNA | AF073489.1 | - | 9 - 10 | 1113 | 119 | |
AK030947.1 | - | 11 - 12 | 2517 | 119 | ||
AK050804.1 | - | 11 - 12 | 1475 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |