Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198029.2 | NP_932146.2 | 9 - 10 | 1870 | 83 | |
GenBank mRNA | AF073489.1 | - | 6 - 7 | 656 | 83 | |
AK030947.1 | - | 8 - 9 | 2060 | 83 | ||
AK050804.1 | - | 8 - 9 | 1018 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |