Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027588.3 | NP_081864.2 | 1 - 2 | 130 | 146 | |
GenBank mRNA | AF356186.1 | - | 1 - 2 | 103 | 146 | |
AK014892.1 | - | 1 - 2 | 130 | 146 | ||
BC060976.1 | - | 1 - 2 | 122 | 6773248 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |