Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134040.1 | NP_598801.1 | 11 - 12 | 758 | 150 | |
GenBank mRNA | AK028341.1 | - | 11 - 12 | 755 | 150 | |
AK132157.1 | - | 11 - 12 | 798 | 150 | ||
AK153335.1 | - | 11 - 12 | 782 | 150 | ||
AK160982.1 | - | 11 - 12 | 759 | 150 | ||
AK166146.1 | - | 11 - 12 | 152 | |||
BC008570.1 | - | 11 - 12 | 736 | 3587491 | 150 | |
BC010624.1 | - | 11 - 12 | 758 | 3153029 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |