Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134040.1 | NP_598801.1 | 8 - 9 | 534 | 63 | |
GenBank mRNA | AK028341.1 | - | 8 - 9 | 531 | 63 | |
AK153335.1 | - | 8 - 9 | 558 | 63 | ||
AK160982.1 | - | 8 - 9 | 535 | 63 | ||
AK166146.1 | - | 8 - 9 | 514 | 63 | ||
AK217868.1 | - | 2 - 3 | 150 | 63 | ||
BC010624.1 | - | 8 - 9 | 534 | 3153029 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |