Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145529.3 | NP_663504.1 | 10 - 11 | 1021 | 62 | |
GenBank mRNA | AK159180.1 | - | 10 - 11 | 824 | 62 | |
BC003241.1 | - | 10 - 11 | 982 | 3498424 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |