Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008421.1 | NP_001008421.1 | 19 - 20 | 77 | ||
GenBank mRNA | AK148210.1 | - | 19 - 20 | 1921 | 77 | |
AK160864.1 | - | 19 - 20 | 77 | |||
AK166160.1 | - | 19 - 20 | 77 | |||
BC086676.1 | - | 19 - 20 | 30659737 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |