Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008421.1 | NP_001008421.1 | 20 - 21 | 2009 | 65 | |
GenBank mRNA | AK148210.1 | - | 20 - 21 | 2025 | 65 | |
AK160864.1 | - | 20 - 21 | 2045 | 65 | ||
AK166160.1 | - | 20 - 21 | 2039 | 65 | ||
BC086676.1 | - | 20 - 21 | 2009 | 30659737 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |