Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172856.3 | NP_766444.1 | 3 - 4 | 480 | 86 | |
GenBank mRNA | AK028849.1 | - | 3 - 4 | 481 | 86 | |
AK135798.1 | - | 3 - 4 | 582 | 86 | ||
AK151848.1 | - | 3 - 4 | 569 | 86 | ||
AK152417.1 | - | 3 - 4 | 550 | 86 | ||
BC057629.1 | - | 3 - 4 | 523 | 4224547 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |