Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172856.3 | NP_766444.1 | 4 - 5 | 551 | 84 | |
GenBank mRNA | AK028849.1 | - | 4 - 5 | 552 | 84 | |
AK135798.1 | - | 4 - 5 | 653 | 84 | ||
AK151848.1 | - | 4 - 5 | 640 | 84 | ||
AK152417.1 | - | 4 - 5 | 621 | 84 | ||
BC057629.1 | - | 4 - 5 | 594 | 4224547 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |