Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025314.3 | NP_079590.1 | 2 - 3 | 237 | 66 | |
GenBank mRNA | AK002246.1 | - | 2 - 3 | 173 | 66 | |
AK003594.1 | - | 2 - 3 | 209 | 66 | ||
AK010822.1 | - | 2 - 3 | 175 | 66 | ||
AK011917.1 | - | 2 - 3 | 205 | 66 | ||
AK017565.1 | - | 2 - 3 | 161 | 66 | ||
AK158898.1 | - | 2 - 3 | 294 | 66 | ||
BC026537.1 | - | 2 - 3 | 210 | 5008303 | 66 | |
BY294360.1 | - | 2 - 3 | 237 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |