Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200013581.mm.1 |
NM_145378.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013581.mm.1 |
NR_104353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013581.mm.1 |
TC0200001788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013577.mm.1 |
NM_145378.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013577.mm.1 |
NR_104353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013577.mm.1 |
TC0200001788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013580.mm.1 |
NM_145378.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013580.mm.1 |
NR_104353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013580.mm.1 |
TC0200001788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013579.mm.1 |
NM_145378.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013579.mm.1 |
NR_104353.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013579.mm.1 |
TC0200001788.mm.1 |
MTA TC |