Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024856.2 | NP_001020027.1 | 4 - 5 | 722 | 76 | |
NM_001024857.2 | NP_001020028.1 | 3 - 4 | 648 | 76 | ||
NM_080788.3 | NP_542966.2 | 4 - 5 | 476 | 76 | ||
GenBank mRNA | AB046593.2 | - | 3 - 4 | 400 | 76 | |
AK046978.1 | - | 4 - 5 | 695 | 76 | ||
AK053820.1 | - | 4 - 5 | 711 | 76 | ||
AK077011.1 | - | 4 - 5 | 497 | 76 | ||
AK137024.1 | - | 3 - 4 | 637 | 76 | ||
AK148269.1 | - | 3 - 4 | 652 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |