Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204973.1 | NP_001191902.1 | 7 - 8 | 2911 | 67 | |
NM_010238.3 | NP_034368.2 | 7 - 8 | 2730 | 67 | ||
NR_037970.1 | 7 - 8 | 2557 | 67 | |||
GenBank mRNA | AB010246.1 | - | 6 - 7 | 1235 | 67 | |
AB010247.1 | - | 6 - 7 | 1204 | 67 | ||
AB010248.1 | - | 5 - 6 | 1101 | 67 | ||
AB212273.1 | - | 5 - 6 | 1065 | 67 | ||
AF045462.1 | - | 6 - 7 | 1876 | 67 | ||
AF318183.1 | - | 6 - 7 | 1893 | 67 | ||
AK147918.1 | - | 7 - 8 | 4260 | 67 | ||
AK158970.1 | - | 7 - 8 | 2731 | 67 | ||
AK168423.1 | - | 7 - 8 | 1989 | 67 | ||
AK168525.1 | - | 7 - 8 | 2602 | 67 | ||
AK220444.1 | - | 6 - 7 | 1878 | 67 | ||
BC046809.1 | - | 7 - 8 | 1610 | 67 | ||
BC085251.1 | - | 8 - 9 | 2753 | 67 | ||
BC138655.1 | - | 6 - 7 | 1504 | 8861675 | 67 | |
BC138656.1 | - | 6 - 7 | 1504 | 8861676 | 67 | |
BM947875.1 | - | 1 - 2 | 358 | 67 | ||
KJ650222.1 | - | 6 - 7 | 1196 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |