Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025371.2 | NP_001020542.1 | 1 - 2 | 308 | 76 | |
GenBank mRNA | AK044479.1 | - | 1 - 2 | 335 | 76 | |
BC026459.1 | - | 1 - 2 | 237 | 76 | ||
BC083132.1 | - | 1 - 2 | 308 | 6466398 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |