Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082960.1 | NP_001076429.1 | 15 - 16 | 1945 | 83 | |
NM_008401.2 | NP_032427.2 | 15 - 16 | 1942 | 83 | ||
GenBank mRNA | AK040921.1 | - | 5 - 6 | 670 | 83 | |
AK089521.1 | - | 15 - 16 | 1908 | 83 | ||
AK146569.1 | - | 7 - 8 | 847 | 83 | ||
AK154224.1 | - | 15 - 16 | 1862 | 83 | ||
AK154474.1 | - | 15 - 16 | 1896 | 83 | ||
AK154798.1 | - | 15 - 16 | 83 | |||
AK155006.1 | - | 15 - 16 | 1874 | 83 | ||
AK155520.1 | - | 15 - 16 | 1867 | 83 | ||
AK155671.1 | - | 15 - 16 | 1911 | 83 | ||
AK155717.1 | - | 15 - 16 | 1900 | 83 | ||
AK170324.1 | - | 15 - 16 | 1914 | 83 | ||
AK171068.1 | - | 12 - 13 | 1574 | 83 | ||
AK171150.1 | - | 15 - 16 | 1880 | 83 | ||
EF101557.1 | - | 1 - 2 | 140 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |