Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166584.1 | NP_001160056.1 | 11 - 12 | 1410 | 65 | |
NM_001166585.1 | NP_001160057.1 | 8 - 9 | 928 | 65 | ||
NM_009346.3 | NP_033372.1 | 10 - 11 | 1347 | 65 | ||
GenBank mRNA | AK140307.1 | - | 11 - 12 | 1411 | 65 | |
AK148375.1 | - | 10 - 11 | 1313 | 65 | ||
BC052021.1 | - | 8 - 9 | 869 | 65 | ||
BC060138.1 | - | 8 - 9 | 869 | 6410573 | 65 | |
L06865.1 | - | 8 - 9 | 874 | 65 | ||
L13853.1 | - | 10 - 11 | 1074 | 65 | ||
S74227.1 | - | 9 - 10 | 1335 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |