Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130452.1 | NP_569719.1 | 4 - 5 | 738 | 62 | |
GenBank mRNA | AK050010.1 | - | 756 | 62 | ||
AY033514.1 | - | 4 - 5 | 676 | 62 | ||
BC019406.1 | - | 4 - 5 | 738 | 5134018 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |