Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008653.2 | NP_032679.2 | 18 - 19 | 1826 | 77 | |
GenBank mRNA | AF097333.1 | - | 18 - 19 | 1777 | 77 | |
AK146661.1 | - | 18 - 19 | 1827 | 77 | ||
AK146886.1 | - | 18 - 19 | 1827 | 77 | ||
AK169302.1 | - | 17 - 18 | 1804 | 77 | ||
AK169306.1 | - | 17 - 18 | 1804 | 77 | ||
BC094945.1 | - | 18 - 19 | 1823 | 77 | ||
BC131672.1 | - | 18 - 19 | 1802 | 77 | ||
BC154408.1 | - | 18 - 19 | 1847 | 40110890 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |