Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029170.1 | - | 982 | 100 | ||
AK122489.1 | - | 1 - 2 | 3031 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |