Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccndbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010761.2 | NP_034891.2 | 7 - 8 | 667 | 77 | |
GenBank mRNA | AK144883.1 | - | 7 - 8 | 644 | 77 | |
AK154285.1 | - | 7 - 8 | 621 | 77 | ||
AK160205.1 | - | 7 - 8 | 633 | 77 | ||
AK163150.1 | - | 7 - 8 | 580 | 77 | ||
AK167345.1 | - | 7 - 8 | 632 | 77 | ||
BC001984.1 | - | 7 - 8 | 603 | 3489973 | 77 | |
BC004673.1 | - | 1 - 2 | 87 | 77 | ||
BC005802.1 | - | 6 - 7 | 676 | 3707919 | 77 | |
U50734.1 | - | 7 - 8 | 585 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |