Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024432.2 | NP_077752.1 | 8 - 9 | 989 | 85 | |
GenBank mRNA | AF272895.1 | - | 8 - 9 | 1004 | 85 | |
AK008942.1 | - | 8 - 9 | 997 | 85 | ||
AK009117.1 | - | 7 - 8 | 771 | 85 | ||
AK075705.1 | - | 6 - 7 | 2053 | 85 | ||
AK082510.1 | - | 1028 | 85 | |||
AK131187.1 | - | 2 - 3 | 2761 | 85 | ||
AK148170.1 | - | 957 | 85 | |||
AK161273.1 | - | 8 - 9 | 1010 | 85 | ||
BC016412.1 | - | 8 - 9 | 918 | 4215883 | 85 | |
BC049963.1 | - | 8 - 9 | 989 | 2655935 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |