Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyltl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166633.2 | NP_001160105.1 | 12 - 13 | 1695 | 69 | |
NM_001290773.1 | NP_001277702.1 | 13 - 14 | 1803 | 69 | ||
NM_001290774.1 | NP_001277703.1 | 12 - 13 | 1717 | 69 | ||
NM_001290775.1 | NP_001277704.1 | 13 - 14 | 2020 | 69 | ||
NM_172670.3 | NP_766258.2 | 13 - 14 | 1800 | 69 | ||
GenBank mRNA | AK077630.1 | - | 12 - 13 | 1671 | 69 | |
AY742914.1 | - | 13 - 14 | 1776 | 69 | ||
AY742915.1 | - | 12 - 13 | 1671 | 69 | ||
BC004023.1 | - | 2 - 3 | 405 | 69 | ||
BC033922.1 | - | 3 - 4 | 453 | 69 | ||
BC058199.1 | - | 13 - 14 | 1981 | 69 | ||
BC096655.1 | - | 12 - 13 | 1727 | 5057489 | 69 | |
BC116715.1 | - | 12 - 13 | 1732 | 40093557 | 69 | |
HM141800.1 | - | 12 - 13 | 1629 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |