Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027921.1 | NP_082197.1 | 1 - 2 | 220 | 94 | |
GenBank mRNA | BC023456.1 | - | 1 - 2 | 220 | 5041164 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |