Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024181.2 | NP_077143.2 | 19 - 20 | 2477 | 121 | |
GenBank mRNA | AK004617.1 | - | 19 - 20 | 2232 | 121 | |
AK028184.1 | - | 19 - 20 | 2218 | 121 | ||
AK128945.1 | - | 19 - 20 | 2231 | 121 | ||
AK165652.1 | - | 19 - 20 | 2234 | 121 | ||
AK166502.1 | - | 19 - 20 | 2188 | 121 | ||
AK169020.1 | - | 19 - 20 | 2244 | 121 | ||
BC002207.1 | - | 10 - 11 | 1039 | 121 | ||
BC033461.1 | - | 19 - 20 | 2216 | 4010341 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |