Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010806.1 | NP_034936.1 | 6 - 7 | 1032 | 107 | |
GenBank mRNA | AF172447.1 | - | 6 - 7 | 1031 | 107 | |
AK016557.1 | - | 6 - 7 | 1196 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |