Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178635.3 | NP_848750.3 | 4 - 5 | 629 | 83 | |
GenBank mRNA | AB020334.1 | - | 3 - 4 | 234 | 83 | |
AK030814.1 | - | 4 - 5 | 560 | 83 | ||
AK043084.1 | - | 4 - 5 | 607 | 83 | ||
AK078085.1 | - | 4 - 5 | 621 | 83 | ||
AK089900.1 | - | 4 - 5 | 614 | 83 | ||
AK154903.1 | - | 4 - 5 | 629 | 83 | ||
AK165015.1 | - | 4 - 5 | 617 | 83 | ||
AK165185.1 | - | 4 - 5 | 609 | 83 | ||
BC034176.1 | - | 4 - 5 | 598 | 4954871 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |