Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013737.5 | NP_038765.2 | 6 - 7 | 913 | 76 | |
GenBank mRNA | AK005158.1 | - | 6 - 7 | 650 | 76 | |
AK005210.1 | - | 6 - 7 | 707 | 76 | ||
AK051454.1 | - | 6 - 7 | 914 | 76 | ||
AK155678.1 | - | 7 - 8 | 764 | 76 | ||
BC010726.1 | - | 6 - 7 | 669 | 3707891 | 76 | |
U34277.1 | - | 6 - 7 | 659 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |