Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009358.3 | NP_033384.2 | 12 - 13 | 1599 | 96 | |
GenBank mRNA | AB053464.1 | - | 12 - 13 | 1544 | 96 | |
AF316616.2 | - | 12 - 13 | 1456 | 96 | ||
AK160587.1 | - | 12 - 13 | 1540 | 96 | ||
AK204058.1 | - | 2 - 3 | 225 | 96 | ||
AY338234.1 | - | 12 - 13 | 1529 | 96 | ||
BC009163.1 | - | 2 - 3 | 126 | 96 | ||
BC010716.1 | - | 12 - 13 | 1512 | 2655861 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |