Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028751.3 | NP_083027.1 | 2 - 3 | 163 | 85 | |
GenBank mRNA | AK002882.2 | - | 2 - 3 | 150 | 85 | |
AK190015.1 | - | 1 - 2 | 58 | 85 | ||
BC023316.1 | - | 2 - 3 | 149 | 3993257 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |