Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025878.1 | NP_080154.1 | 2 - 3 | 187 | 62 | |
GenBank mRNA | AB049954.1 | - | 2 - 3 | 211 | 62 | |
AK010250.1 | - | 2 - 3 | 194 | 62 | ||
AK088540.1 | - | 2 - 3 | 183 | 62 | ||
AK166597.1 | - | 2 - 3 | 208 | 62 | ||
AK167067.1 | - | 2 - 3 | 188 | 62 | ||
AK168140.1 | - | 2 - 3 | 203 | 62 | ||
BC021752.1 | - | 2 - 3 | 187 | 5136720 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |