Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146211.1 | NP_001139683.1 | 4 - 5 | 347 | 83 | |
NM_001146212.1 | NP_001139684.1 | 4 - 5 | 350 | 83 | ||
NM_183086.2 | NP_898909.2 | 4 - 5 | 440 | 83 | ||
GenBank mRNA | AK004151.1 | - | 4 - 5 | 344 | 83 | |
BB862036.1 | - | 4 - 5 | 359 | 83 | ||
BC038917.1 | - | 4 - 5 | 336 | 83 | ||
BC048912.1 | - | 4 - 5 | 443 | 4238014 | 83 | |
BC096043.1 | - | 3 - 4 | 305 | 83 | ||
BC096657.1 | - | 3 - 4 | 255 | 5144028 | 83 | |
DV046794.1 | - | 4 - 5 | 361 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |